Learn More
Cell Signaling Technology PTMScan™ Acetyl-Lysine Motif Kit, Agarose, 10 Assays
SDP
Catalog No. 503131507
Encompass
The PTMScan™ Acetyl-Lysine Motif [Ac-K] Kit enables the quantitative profiling of acetylated lysine sites in cellular proteins. Utilizing a proprietary antibody-based enrichment methodology, this kit allows for the isolation and identification of post-translationally modified peptides from complex samples. When combined with liquid chromatography-tandem mass spectrometry (LC-MS/MS), it provides a global and unbiased overview of acetylation status across various cell and tissue types.
  • Enables quantitative profiling of acetylated lysine sites
  • Proprietary bead-conjugated antibody facilitates specific peptide enrichment
  • Compatible with LC-MS/MS workflows for large-scale proteomic analysis
  • Supports unbiased discovery of post-translational modifications
  • Includes immunoaffinity beads and optimized buffer for 10 assays
Catalog No. 50-313-1507 Supplier Cell Signaling Technology Supplier No. 13416S
The PTMScan™ Acetyl-Lysine Motif [Ac-K] Kit enables the quantitative profiling of acetylated lysine sites in cellular proteins. Utilizing a proprietary antibody-based enrichment methodology, this kit allows for the isolation and identification of post-translationally modified peptides from complex samples. When combined with liquid chromatography-tandem mass spectrometry (LC-MS/MS), it provides a global and unbiased overview of acetylation status across various cell and tissue types.
  • Enables quantitative profiling of acetylated lysine sites
  • Proprietary bead-conjugated antibody facilitates specific peptide enrichment
  • Compatible with LC-MS/MS workflows for large-scale proteomic analysis
  • Supports unbiased discovery of post-translational modifications
  • Includes immunoaffinity beads and optimized buffer for 10 assays