Learn More
Cell Signaling Technology PTMScan™ Asymmetric Di-Methyl Arginine Motif Kit, 10 Assays
SDP
Catalog No. 503133544
Encompass
The PTMScan™ Asymmetric Di-Methyl Arginine Motif [adme-R] Kit employs a proprietary methodology for peptide enrichment by immunoprecipitation. It utilizes a specific bead-conjugated antibody in combination with liquid chromatography tandem mass spectrometry (LC-MS/MS) for the quantitative profiling of asymmetric dimethylarginine sites in cellular proteins. This kit allows researchers to isolate, identify, and quantitate post-translationally modified cellular peptides with high specificity and sensitivity.
  • Enables global, unbiased profiling of post-translational modification sites
  • Designed for peptide enrichment and mass spectrometry analysis
  • Features antibody conjugated to protein A agarose beads
  • Validated for use in research applications
  • Supplied with immunoaffinity beads and IAP buffer
Catalog No. 50-313-3544 Supplier Cell Signaling Technology Supplier No. 13474S
The PTMScan™ Asymmetric Di-Methyl Arginine Motif [adme-R] Kit employs a proprietary methodology for peptide enrichment by immunoprecipitation. It utilizes a specific bead-conjugated antibody in combination with liquid chromatography tandem mass spectrometry (LC-MS/MS) for the quantitative profiling of asymmetric dimethylarginine sites in cellular proteins. This kit allows researchers to isolate, identify, and quantitate post-translationally modified cellular peptides with high specificity and sensitivity.
  • Enables global, unbiased profiling of post-translational modification sites
  • Designed for peptide enrichment and mass spectrometry analysis
  • Features antibody conjugated to protein A agarose beads
  • Validated for use in research applications
  • Supplied with immunoaffinity beads and IAP buffer