Learn More
Cell Signaling Technology PTMScan™ Glutaryl-Lysine Kit, 10 Assays, 1 Kit
SDP
Catalog No. 503132630
Encompass
The PTMScan™ Glutaryl-Lysine [Glut-K] Kit utilizes immunoprecipitation to facilitate the quantitative profiling of glutaryl-lysine post-translational modification sites within cellular proteins. By combining bead-conjugated antibodies with liquid chromatography tandem mass spectrometry (LC-MS/MS), this kit allows researchers to isolate and identify modified peptides with high specificity and sensitivity. It provides a global, unbiased overview of lysine glutarylation across diverse cell and tissue samples.
  • Enables large-scale isolation and quantification of glutarylated peptides
  • Includes 10 assays for peptide enrichment by immunoaffinity purification
  • Supplied with IAP buffer and glutaryl-lysine immunoaffinity beads
  • Compatible with standard LC-MS/MS workflows for proteomic analysis
  • Validated for diverse sample types with global species homology
Catalog No. 50-313-2630 Supplier Cell Signaling Technology Supplier No. 26101S
The PTMScan™ Glutaryl-Lysine [Glut-K] Kit utilizes immunoprecipitation to facilitate the quantitative profiling of glutaryl-lysine post-translational modification sites within cellular proteins. By combining bead-conjugated antibodies with liquid chromatography tandem mass spectrometry (LC-MS/MS), this kit allows researchers to isolate and identify modified peptides with high specificity and sensitivity. It provides a global, unbiased overview of lysine glutarylation across diverse cell and tissue samples.
  • Enables large-scale isolation and quantification of glutarylated peptides
  • Includes 10 assays for peptide enrichment by immunoaffinity purification
  • Supplied with IAP buffer and glutaryl-lysine immunoaffinity beads
  • Compatible with standard LC-MS/MS workflows for proteomic analysis
  • Validated for diverse sample types with global species homology