Learn More
Cell Signaling Technology PTMScan™ HS Acetyl-Lysine Motif Kit, 10 Assays, 1 Kit
SDP
Catalog No. 503131497
Encompass
The PTMScan™ HS (High Sensitivity) Acetyl-Lysine Motif (Ac-K) Kit offers an enhanced methodology for the identification of post-translationally modified peptides. Utilizing a proprietary immunoprecipitation process with magnetic bead-conjugated antibodies and liquid chromatography-tandem mass spectrometry (LC-MS/MS), this kit allows for the quantitative profiling of acetyl-lysine PTM sites in cellular proteins. The workflow provides a global, unbiased overview of protein acetylation across diverse cell and tissue samples.
  • Enables quantitative profiling of PTM sites in cellular proteins.
  • Delivers high specificity and sensitivity for PTM peptide enrichment.
  • Facilitates global, unbiased overview of acetylation across species.
  • Supports 10 assays per kit.
Catalog No. 50-313-1497 Supplier Cell Signaling Technology Supplier No. 46784S
The PTMScan™ HS (High Sensitivity) Acetyl-Lysine Motif (Ac-K) Kit offers an enhanced methodology for the identification of post-translationally modified peptides. Utilizing a proprietary immunoprecipitation process with magnetic bead-conjugated antibodies and liquid chromatography-tandem mass spectrometry (LC-MS/MS), this kit allows for the quantitative profiling of acetyl-lysine PTM sites in cellular proteins. The workflow provides a global, unbiased overview of protein acetylation across diverse cell and tissue samples.
  • Enables quantitative profiling of PTM sites in cellular proteins.
  • Delivers high specificity and sensitivity for PTM peptide enrichment.
  • Facilitates global, unbiased overview of acetylation across species.
  • Supports 10 assays per kit.