Learn More
Cell Signaling Technology PTMScan™ HS Succinyl-Lysine Motif Kit, 10 Assays, 1 Kit
SDP
Catalog No. 503133262
Encompass
The PTMScan™ HS Succinyl-Lysine Motif (Succ-K) Kit provides an enhanced methodology for the enrichment and identification of succinyl-lysine modified peptides. Utilizing proprietary immunoaffinity purification with magnetic bead-conjugated antibodies, this kit enables the quantitative profiling of post-translationally modified sites via liquid chromatography-tandem mass spectrometry (LC-MS/MS). The workflow facilitates a comprehensive and unbiased analysis of lysine succinylation across diverse cell and tissue samples.
  • High-sensitivity enrichment of succinylated peptides for mass spectrometry analysis.
  • Includes magnetic immunoaffinity beads and optimized bind and wash buffers.
  • Supports quantitative profiling of post-translational modifications in complex biological samples.
  • Validated for use in PTMScan applications.
Catalog No. 50-313-3262 Supplier Cell Signaling Technology Supplier No. 60724S
The PTMScan™ HS Succinyl-Lysine Motif (Succ-K) Kit provides an enhanced methodology for the enrichment and identification of succinyl-lysine modified peptides. Utilizing proprietary immunoaffinity purification with magnetic bead-conjugated antibodies, this kit enables the quantitative profiling of post-translationally modified sites via liquid chromatography-tandem mass spectrometry (LC-MS/MS). The workflow facilitates a comprehensive and unbiased analysis of lysine succinylation across diverse cell and tissue samples.
  • High-sensitivity enrichment of succinylated peptides for mass spectrometry analysis.
  • Includes magnetic immunoaffinity beads and optimized bind and wash buffers.
  • Supports quantitative profiling of post-translational modifications in complex biological samples.
  • Validated for use in PTMScan applications.