Learn More
Cell Signaling Technology PTMScan™ Mono-Methyl Lysine Motif Kit, 10 Assays
SDP
Catalog No. 503133021
Encompass
The PTMScan™ Mono-Methyl Lysine Motif (mme-K) Kit utilizes proprietary technology for the enrichment of post-translationally modified peptides via immunoprecipitation, followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) for quantitative profiling. This workflow allows for the global, unbiased identification and quantification of mono-methylated lysine sites in cellular proteins across diverse cell and tissue samples.
  • Enables isolation and quantification of large numbers of post-translationally modified cellular peptides
  • High degree of specificity and sensitivity for PTM site profiling
  • Provides an unbiased, global overview of protein methylation
  • Designed for use with liquid chromatography tandem mass spectrometry
  • Supplied with immunoaffinity beads and optimized buffer for 10 assays
Catalog No. 50-313-3021 Supplier Cell Signaling Technology Supplier No. 16892S
The PTMScan™ Mono-Methyl Lysine Motif (mme-K) Kit utilizes proprietary technology for the enrichment of post-translationally modified peptides via immunoprecipitation, followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) for quantitative profiling. This workflow allows for the global, unbiased identification and quantification of mono-methylated lysine sites in cellular proteins across diverse cell and tissue samples.
  • Enables isolation and quantification of large numbers of post-translationally modified cellular peptides
  • High degree of specificity and sensitivity for PTM site profiling
  • Provides an unbiased, global overview of protein methylation
  • Designed for use with liquid chromatography tandem mass spectrometry
  • Supplied with immunoaffinity beads and optimized buffer for 10 assays