Learn More
Cell Signaling Technology PTMScan Phospho-Ser/Thr Motif Kit, 10 Assays, 1 Kit
SDP
Catalog No. 503133667
Encompass
PTMScan™ technology enables the quantitative profiling of post-translational modification (PTM) sites in cellular proteins through a proprietary methodology. By utilizing a bead-conjugated antibody for immunoprecipitation, this kit facilitates the enrichment of specific peptides, which are subsequently analyzed via liquid chromatography-tandem mass spectrometry (LC-MS/MS). This workflow allows researchers to identify and quantify large numbers of modified cellular peptides across various cell and tissue samples.
  • Enables global, unbiased profiling of post-translational modifications
  • Includes 10 assays for peptide enrichment
  • Validated for use in high-sensitivity quantitative mass spectrometry analysis
  • Designed for specificity using motif-specific antibodies conjugated to agarose beads
  • Requires storage at -20°C
Catalog No. 50-313-3667 Supplier Cell Signaling Technology Supplier No. 25081S
PTMScan™ technology enables the quantitative profiling of post-translational modification (PTM) sites in cellular proteins through a proprietary methodology. By utilizing a bead-conjugated antibody for immunoprecipitation, this kit facilitates the enrichment of specific peptides, which are subsequently analyzed via liquid chromatography-tandem mass spectrometry (LC-MS/MS). This workflow allows researchers to identify and quantify large numbers of modified cellular peptides across various cell and tissue samples.
  • Enables global, unbiased profiling of post-translational modifications
  • Includes 10 assays for peptide enrichment
  • Validated for use in high-sensitivity quantitative mass spectrometry analysis
  • Designed for specificity using motif-specific antibodies conjugated to agarose beads
  • Requires storage at -20°C