Learn More
Cell Signaling Technology PTMScan Propionyl-Lysine Kit, 10 Assays, 1 Kit
SDP
Catalog No. 503133101
Encompass
The PTMScan™ Propionyl-Lysine [Prop-K] Kit utilizes proprietary immunoaffinity enrichment technology to facilitate the quantitative profiling of propionyl-lysine post-translational modification sites within cellular proteins. By combining specific bead-conjugated antibodies with liquid chromatography tandem mass spectrometry (LC-MS/MS), this kit allows researchers to isolate and identify modified peptides with high sensitivity and specificity. The methodology is designed for unbiased, global analysis of cellular and tissue samples.
  • Enables large-scale identification and quantitation of propionyl-lysine modified peptides
  • Includes high-affinity motif antibody beads for robust immunoprecipitation
  • Optimized for downstream LC-MS/MS analysis
  • Supplied with concentrated IAP buffer for consistent experimental results
  • Suitable for 10 assays
Catalog No. 50-313-3101 Supplier Cell Signaling Technology Supplier No. 17848S
The PTMScan™ Propionyl-Lysine [Prop-K] Kit utilizes proprietary immunoaffinity enrichment technology to facilitate the quantitative profiling of propionyl-lysine post-translational modification sites within cellular proteins. By combining specific bead-conjugated antibodies with liquid chromatography tandem mass spectrometry (LC-MS/MS), this kit allows researchers to isolate and identify modified peptides with high sensitivity and specificity. The methodology is designed for unbiased, global analysis of cellular and tissue samples.
  • Enables large-scale identification and quantitation of propionyl-lysine modified peptides
  • Includes high-affinity motif antibody beads for robust immunoprecipitation
  • Optimized for downstream LC-MS/MS analysis
  • Supplied with concentrated IAP buffer for consistent experimental results
  • Suitable for 10 assays