Learn More
Cell Signaling Technology PTMScan Succinyl-Lysine Motif Kit, 10 Assays
SDP
Catalog No. 503131880
Encompass
The PTMScan™ Succinyl-Lysine Motif [Succ-K] Kit utilizes a proprietary immunoprecipitation methodology to enrich post-translationally modified peptides for analysis by liquid chromatography-tandem mass spectrometry (LC-MS/MS). This kit enables researchers to isolate, identify, and quantitate succinylated lysine peptides from cellular and tissue samples, providing a global, unbiased profile of this regulatory modification.
  • Provides high-sensitivity enrichment of succinyl-lysine modified peptides using specific bead-conjugated antibodies.
  • Compatible with diverse biological samples for comprehensive proteomics profiling.
  • Facilitates accurate quantification of PTM sites across numerous protein substrates.
  • Includes high-quality immunoaffinity beads and buffer optimized for peptide capture.
  • Designed for research use in identifying protein modifications in cellular pathways.
Catalog No. 50-313-1880 Supplier Cell Signaling Technology Supplier No. 13764S
The PTMScan™ Succinyl-Lysine Motif [Succ-K] Kit utilizes a proprietary immunoprecipitation methodology to enrich post-translationally modified peptides for analysis by liquid chromatography-tandem mass spectrometry (LC-MS/MS). This kit enables researchers to isolate, identify, and quantitate succinylated lysine peptides from cellular and tissue samples, providing a global, unbiased profile of this regulatory modification.
  • Provides high-sensitivity enrichment of succinyl-lysine modified peptides using specific bead-conjugated antibodies.
  • Compatible with diverse biological samples for comprehensive proteomics profiling.
  • Facilitates accurate quantification of PTM sites across numerous protein substrates.
  • Includes high-quality immunoaffinity beads and buffer optimized for peptide capture.
  • Designed for research use in identifying protein modifications in cellular pathways.