Learn More
Cell Signaling Technology PTMScan™ Pilot Acetyl-Lysine Motif Kit, 3 Assays
SDP
Catalog No. 503133107
Encompass
The PTMScan™ Pilot Acetyl-Lysine Motif [Ac-K] Kit utilizes proprietary immunoaffinity purification technology to enable quantitative profiling of acetylated lysine sites in cellular proteins. By combining agarose bead-conjugated motif antibodies with liquid chromatography-tandem mass spectrometry (LC-MS/MS), the kit facilitates the global, unbiased identification and enrichment of post-translationally modified peptides from protease-digested samples.
  • Enables isolation and quantification of endogenous peptides containing the acetylated lysine motif.
  • Includes immunoaffinity beads and 10X IAP buffer for three assays.
  • Supports high-specificity enrichment of post-translationally modified peptides.
  • Designed for use in downstream mass spectrometry workflows.
Catalog No. 50-313-3107 Supplier Cell Signaling Technology Supplier No. 14499S
The PTMScan™ Pilot Acetyl-Lysine Motif [Ac-K] Kit utilizes proprietary immunoaffinity purification technology to enable quantitative profiling of acetylated lysine sites in cellular proteins. By combining agarose bead-conjugated motif antibodies with liquid chromatography-tandem mass spectrometry (LC-MS/MS), the kit facilitates the global, unbiased identification and enrichment of post-translationally modified peptides from protease-digested samples.
  • Enables isolation and quantification of endogenous peptides containing the acetylated lysine motif.
  • Includes immunoaffinity beads and 10X IAP buffer for three assays.
  • Supports high-specificity enrichment of post-translationally modified peptides.
  • Designed for use in downstream mass spectrometry workflows.