Learn More
Aviva Systems Biology Corp PTN Antibody - C-terminal region (ARP45356_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45356_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PTN. Alternate names: HARP, HBGF8, HBNF, NEGF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59011