Learn More
SANISURE Pharma-Clear R Braided Silicone 3/8" ID
Catalog No. NC2165654
Encompass
Pharma-Clear R platinum-cured-silicone braided hose perfect for pharmaceutical and bio-processing and is also commonly used for high-purity chemicals and food and beverage applications. Our proprietary polyester yarn inner braid was engineered to achieve higher pressures and maximum consistency when compared to other brands of braided silicone hose. The inner bore of Pharma-Clear R is manufactured using the same compound as our PLTS-65 series tubing which can be valuable for those looking to minimize the number of process-contact materials. Download our comprehensive tubing brochure today to learn more.
This item is not returnable. View return policy
Catalog No. NC2165654 Supplier SANISURE Supplier No. PLTSR37550
Pharma-Clear R platinum-cured-silicone braided hose perfect for pharmaceutical and bio-processing and is also commonly used for high-purity chemicals and food and beverage applications. Our proprietary polyester yarn inner braid was engineered to achieve higher pressures and maximum consistency when compared to other brands of braided silicone hose. The inner bore of Pharma-Clear R is manufactured using the same compound as our PLTS-65 series tubing which can be valuable for those looking to minimize the number of process-contact materials. Download our comprehensive tubing brochure today to learn more.