Learn More
Aviva Systems Biology Corp PTOV1 Antibody - N-terminal region (ARP50614_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50614_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTOV1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PTOV1. Alternate names: DKFZP586I111, MGC71475, ACID2, PTOV-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-53637