Learn More
Eurofins Discoverx PTP-1B, 250 µg. (14-621M)
Supplier:  Eurofins Discoverx 14621M
Encompass
The PTP-1B product is purified N terminal GST-tagged recombinant PTP-1B enzyme. This enzyme contains amino acids 1-321. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 0.63 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9070