Learn More
Medchemexpress LLC PTP Inhibitor 1 | 2632-13-5 | 99.55% | 229.07 | 10 G
SDP
Catalog No. 5000454274
Encompass_Preferred
PTP inhibitor 1 is a protein tyrosine phosphatase (PTP) inhibitor that exhibits an anti-angiogenic effect. It is intended for research use only.
  • Inhibits PTP.
  • Shows anti-angiogenic activity with an IC50 of 3.7 μM for HUVECs.
Catalog No. 50-004-54274 Supplier Medchemexpress LLC Supplier No. HYW01347810G
PTP inhibitor 1 is a protein tyrosine phosphatase (PTP) inhibitor that exhibits an anti-angiogenic effect. It is intended for research use only.
  • Inhibits PTP.
  • Shows anti-angiogenic activity with an IC50 of 3.7 μM for HUVECs.