Learn More
Apexbio Technology LLC PTP Inhibitor I 2491-38-5 10g
SDP
Catalog No. 5000089581
Encompass_Preferred
PTP Inhibitor I (CAS 2491-38-5) is a small molecule inhibitor targeting protein tyrosine phosphatases (PTPs) specifically SHP-1 and PTP1B with inhibition constants (KI) of 43 M and 42 M respectively As an -bromoacetophenone derivative it acts as a neutral phosphotyrosine mimetic covalently modifying the active site cysteine of PTPs Structure-activity studies indicate that modifications to the phenyl ring moderately affect potency while para-substituted peptidyl groups can enhance both potency and selectivity PTP Inhibitor I is utilized in vitro for mechanistic studies of PTP function related to diabetes neurodegenerative disorders immune regulation and pathogen virulence Animal and clinical data are not available
Catalog No. 50-000-89581 Supplier Apexbio Technology LLC Supplier No. C360710000
PTP Inhibitor I (CAS 2491-38-5) is a small molecule inhibitor targeting protein tyrosine phosphatases (PTPs) specifically SHP-1 and PTP1B with inhibition constants (KI) of 43 M and 42 M respectively As an -bromoacetophenone derivative it acts as a neutral phosphotyrosine mimetic covalently modifying the active site cysteine of PTPs Structure-activity studies indicate that modifications to the phenyl ring moderately affect potency while para-substituted peptidyl groups can enhance both potency and selectivity PTP Inhibitor I is utilized in vitro for mechanistic studies of PTP function related to diabetes neurodegenerative disorders immune regulation and pathogen virulence Animal and clinical data are not available