Learn More
Selleck Chemical LLC PTP Inhibitor I S3696-25mg
SDP
Supplier:  Selleck Chemical LLC S369625MG
Encompass_Preferred
PTP Inhibitor I is a cell-permeable protein tyrosine phosphatase (PTP) inhibitor with Ki values of 43 and 42 M for SHP-1( SH2) and PTP1B respectively
Catalog No. 50-310-8760