Learn More
Selleck Chemical LLC PTP Inhibitor II S3678-25mg
SDP
Catalog No. 503110749
Encompass_Preferred
PTP Inhibitor II (NSC 129010 4-(Bromoacetyl)anisole) is a cell-permeable protein tyrosine phosphatase (PTP) inhibitor that covalently binds the catalytic domain of the Src homology region 2 domain-containing phosphatase (SHP-1( SH2)) with Ki value of 128 M
Catalog No. 50-311-0749 Supplier Selleck Chemical LLC Supplier No. S367825MG
Add to Cart
Add to Cart
PTP Inhibitor II (NSC 129010 4-(Bromoacetyl)anisole) is a cell-permeable protein tyrosine phosphatase (PTP) inhibitor that covalently binds the catalytic domain of the Src homology region 2 domain-containing phosphatase (SHP-1( SH2)) with Ki value of 128 M