Learn More
Cayman Chemical PTP Inhibitor I, Purity: ≥98%, CAS: 2491-38-5, 50 mg
SDP
Catalog No. 502985467
Encompass_Preferred
A SHP-1 and PTP1B inhibitor (Kis = 43 and 42 µM for the SHP-1 catalytic domain and full-length PTP1B, respectively); inhibits endothelial cell network formation of HUVECs cultured with normal human dermal fibroblasts (IC50 = 3.7 µM); has been used in the synthesis of compounds with antibacterial, antifungal, fungicidal, and anticancer activities
Catalog No. 50-298-5467 Supplier Cayman Chemical Supplier No. 1976650 MG
A SHP-1 and PTP1B inhibitor (Kis = 43 and 42 µM for the SHP-1 catalytic domain and full-length PTP1B, respectively); inhibits endothelial cell network formation of HUVECs cultured with normal human dermal fibroblasts (IC50 = 3.7 µM); has been used in the synthesis of compounds with antibacterial, antifungal, fungicidal, and anticancer activities