Learn More
Cayman Chemical PTP Inhibitor II, Purity: ≥98%, CAS: 2632-13-5, 100 g
SDP
Catalog No. 502983457
Encompass_Preferred
A cell-permeable inhibitor of the catalytic domain of SHP-1, SHP-1(ΔSH2) (Ki = 128 µM)
Catalog No. 50-298-3457 Supplier Cayman Chemical Supplier No. 20629100 G
A cell-permeable inhibitor of the catalytic domain of SHP-1, SHP-1(ΔSH2) (Ki = 128 µM)