Learn More
Cayman Chemical PTP Inhibitor II, Purity: ≥98%, CAS: 2632-13-5, 25 g
SDP
Catalog No. 502985018
Encompass_Preferred
A cell-permeable inhibitor of the catalytic domain of SHP-1, SHP-1(ΔSH2) (Ki = 128 µM)
Catalog No. 50-298-5018 Supplier Cayman Chemical Supplier No. 2062925 G
A cell-permeable inhibitor of the catalytic domain of SHP-1, SHP-1(ΔSH2) (Ki = 128 µM)