Learn More
Cayman Chemical PTP InhIbItr II 50g
SDP
Catalog No. 502981947
Encompass_Preferred
A cell-permeable inhibitor of the catalytic domain of SHP-1, SHP-1(ΔSH2) (Ki = 128 µM)
Catalog No. 50-298-1947 Supplier Cayman Chemical Supplier No. 2062950 G
Add to Cart
Add to Cart
A cell-permeable inhibitor of the catalytic domain of SHP-1, SHP-1(ΔSH2) (Ki = 128 µM)