Learn More
Cayman Chemical PTP Inhibitor III, Purity: ≥95%, CAS: 29936-81-0, 5 mg
SDP
Catalog No. 502983216
Encompass_Preferred
A photoreversible covalent inhibitor of PTPs; binds the catalytic domain of SHP-1 (Ki = 184 µM) and covalently reacts with free thiols, a reaction that is reversible with irradiation (350 nm)
Catalog No. 50-298-3216 Supplier Cayman Chemical Supplier No. 190595 MG
A photoreversible covalent inhibitor of PTPs; binds the catalytic domain of SHP-1 (Ki = 184 µM) and covalently reacts with free thiols, a reaction that is reversible with irradiation (350 nm)