Learn More
Cayman Chemical PTP Inhibitor IV, Purity: ≥98%, CAS: 329317-98-8, 10 mg
SDP
Catalog No. 502979539
Encompass_Preferred
An uncharged, 1,4-di-substituted, phenyl-linked bis-TFMS phosphate mimetic that acts as a reversible, competitive, and active-site directed inhibitor of SHP-2, PTP1B, PTP-ε, PTP-Meg-2, PTP-σ, PTP-β, and PTP-µ (IC50s = 1.8, 2.5, 8.4, 13, 20, 6.4, and 6.7 µM, respectively)
Catalog No. 50-297-9539 Supplier Cayman Chemical Supplier No. 1845810 MG
An uncharged, 1,4-di-substituted, phenyl-linked bis-TFMS phosphate mimetic that acts as a reversible, competitive, and active-site directed inhibitor of SHP-2, PTP1B, PTP-ε, PTP-Meg-2, PTP-σ, PTP-β, and PTP-µ (IC50s = 1.8, 2.5, 8.4, 13, 20, 6.4, and 6.7 µM, respectively)