Learn More
Perkin Elmer US LLC PTP Standard for Elemental Analysis of Oils
Catalog No. 502113673
Encompass_Preferred
Proficiency Testing Program (PTP) was specially designed to give analytical laboratories instant validation while measuring metals, sulfur, viscosity, and particle count in new or used oils. It has been designed to provide laboratories with a method of monitoring their analytical performance as measured against Certified Reference Materials (CRMs): Analyze a sample Report the results online Receive an instant response to submitted values Confirm the accuracy of your analytical results, quickly, easily and definitively. Volume: Content: 25 g PTP Standard
Catalog No. 50-211-3673 Supplier Perkin Elmer US LLC Supplier No. N9308314
Proficiency Testing Program (PTP) was specially designed to give analytical laboratories instant validation while measuring metals, sulfur, viscosity, and particle count in new or used oils. It has been designed to provide laboratories with a method of monitoring their analytical performance as measured against Certified Reference Materials (CRMs): Analyze a sample Report the results online Receive an instant response to submitted values Confirm the accuracy of your analytical results, quickly, easily and definitively. Volume: Content: 25 g PTP Standard