Learn More
Enzo Life Sciences PTP1B 2x Assay Buffer (20ml)
SDP
Catalog No. 502011092
Encompass_Preferred
Formulation: Liquid. In 100 mM HEPES, pH 7.2, containing 2 mM EDTA, 2 mM DTT, 0.1% NP-40. Long Term Storage: -80°C.
Catalog No. 50-201-1092 Supplier Enzo Life Sciences Supplier No. BMLKI1310020
Formulation: Liquid. In 100 mM HEPES, pH 7.2, containing 2 mM EDTA, 2 mM DTT, 0.1% NP-40. Long Term Storage: -80°C.