Learn More
ECM Bioscience PTP1B (a.a.146-157) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP2351
Encompass_Preferred
This antibody was affinity purified with PTP1B (a.a. 146-157) peptide. The purified antibody detects a 50 kDa* band corresponding to PTP1B in western blots of human A431, Jurkat, HeLa, and mouse SYF cells. This reactivity can be specifically blocked using PTP1B (a.a. 146-157) peptide (PX2355). The immunogen was PTP1B synthetic peptide (coupled to carrier protein) corresponds to amino acids 146 to 157 in human PTP1B. This sequence has significant homology to the conserved site in rat and mouse PTP1B, and has homology to TC-PTP and MPTP, but not other protein tyrosine phosphatase family members. Accession #P18031. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PTPN1; TCPTP/PTPN2
Catalog No. 50-172-0871