Learn More
Medchemexpress LLC HY-100462 5mg , PTP1B-IN-2 CAS:1919853-46-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004625MG
Encompass_Preferred
HY-100462 5mg PTP1B-IN-2 CAS: 1919853-46-5 PTP1B-IN-2 is a potent protein tyrosine phosphatase 1B (PTP1B) inhibitor with an IC50 of 50 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1610