Learn More
Medchemexpress LLC PTP1B-IN-3 | 809272-64-8 | C₁₂H₉BrF₂NOP | 5 MG
SDP
Catalog No. 5000374996
Encompass_Preferred
PTP1B-IN-3 is a potent and orally active PTP1B inhibitor with IC50s of 120 nM for both PTP1B and TCPTP. It has antidiabetic and anticancer effects.
  • Potent and orally active PTP1B inhibitor
  • IC50 of 120 nM for PTP1B and TCPTP
  • Exhibits antidiabetic effects
  • Shows anticancer effects
  • Good oral bioavailability in diet-induced obese (DIO) mice
  • Slow clearance and good elimination half-life
Catalog No. 50-003-74996 Supplier Medchemexpress LLC Supplier No. HY151335MG
PTP1B-IN-3 is a potent and orally active PTP1B inhibitor with IC50s of 120 nM for both PTP1B and TCPTP. It has antidiabetic and anticancer effects.
  • Potent and orally active PTP1B inhibitor
  • IC50 of 120 nM for PTP1B and TCPTP
  • Exhibits antidiabetic effects
  • Shows anticancer effects
  • Good oral bioavailability in diet-induced obese (DIO) mice
  • Slow clearance and good elimination half-life