Learn More
Medchemexpress LLC PTP1B-IN-3 Diammonium | 2026852-51-7 | C20H28N4O10P2 | 1 ML
SDP
Catalog No. 5000375805
Encompass_Preferred
PTP1B-IN-3 Diammonium is a potent and selective inhibitor of PTP1B, demonstrating an IC50 of 0.49 nM. It exhibits good selectivity against TCPTP with an IC50 of 330 nM and SHP-2 with an IC50 of 13 μM. This compound has been shown to promote insulin signaling and effectively lower glucose levels in ob/ob mice, making it a valuable tool for research into type 2 diabetes and obesity.
  • Potent and selective PTP1B inhibitor
  • Shows good selectivity against TCPTP and SHP-2
  • Promotes insulin signaling
  • Lowers glucose levels in ob/ob mice
  • Useful for research of type 2 diabetes
  • Useful for research of obesity
Catalog No. 50-003-75805 Supplier Medchemexpress LLC Supplier No. HY15133A1MG
PTP1B-IN-3 Diammonium is a potent and selective inhibitor of PTP1B, demonstrating an IC50 of 0.49 nM. It exhibits good selectivity against TCPTP with an IC50 of 330 nM and SHP-2 with an IC50 of 13 μM. This compound has been shown to promote insulin signaling and effectively lower glucose levels in ob/ob mice, making it a valuable tool for research into type 2 diabetes and obesity.
  • Potent and selective PTP1B inhibitor
  • Shows good selectivity against TCPTP and SHP-2
  • Promotes insulin signaling
  • Lowers glucose levels in ob/ob mice
  • Useful for research of type 2 diabetes
  • Useful for research of obesity