Learn More
Medchemexpress LLC PTP1B-IN-4 | 765317-72-4 | 99.6% | 741.45 g/mol | C26H19Br2N3O7S3 | 10 MG
SDP
Catalog No. 5000287945
Encompass_Preferred
PTP1B-IN-4 is a small-molecule, non-competitive allosteric inhibitor of protein tyrosine phosphatase 1B (PTP1B) with a reported IC50 of approximately 8 μM. It is intended for biochemical and cellular research into PTP1B-related signaling pathways, including studies of metabolism, obesity, and diabetes. The compound is supplied as a powder or as a DMSO solution for research use.
  • Non-competitive allosteric inhibitor of PTP1B (IC50 ≈ 8 μM).
  • High purity (99.55%).
  • Available as powder and as 10 mM solution in DMSO.
  • Pack sizes suitable for small-scale research (1-100 mg).
  • Recommended storage: powder at -20°C; in solvent at -80°C (long term).
Catalog No. 50-002-87945 Supplier Medchemexpress LLC Supplier No. HY1575610MG
PTP1B-IN-4 is a small-molecule, non-competitive allosteric inhibitor of protein tyrosine phosphatase 1B (PTP1B) with a reported IC50 of approximately 8 μM. It is intended for biochemical and cellular research into PTP1B-related signaling pathways, including studies of metabolism, obesity, and diabetes. The compound is supplied as a powder or as a DMSO solution for research use.
  • Non-competitive allosteric inhibitor of PTP1B (IC50 ≈ 8 μM).
  • High purity (99.55%).
  • Available as powder and as 10 mM solution in DMSO.
  • Pack sizes suitable for small-scale research (1-100 mg).
  • Recommended storage: powder at -20°C; in solvent at -80°C (long term).