Learn More
ECM Bioscience PTP1B Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience PK6290
Encompass_Preferred
The PTP1B phospho-regulation antibody sampler kit can be used to detect phosphorylation of PTP1B at Ser-50 and Tyr-152. The kit also includes an antibody to examine total PTP1B expression levels and secondary reagents for rabbit polyclonal and mouse monoclonal antibody detection. Applications: WB, E; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8776