Learn More
Aviva Systems Biology Corp PTP4A3 Antibody - middle region (ARP51193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTP4A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTP4A3. Alternate names: PRL-3, PRL-R, PRL3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53650