Learn More
Eurofins Discoverx PTPβ, active, 250 µg. (14-948M)
Supplier:  Eurofins Discoverx 14948M
Encompass
The PTPβ, active product is purified N terminal GST-tagged recombinant PTPβ enzyme. This enzyme contains amino acids 1643-end. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 0.538 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9467