Learn More
Eurofins Discoverx PTPMEG-1, 250 µg. (14-642M)
Supplier:  Eurofins Discoverx 14642M
Encompass
The PTPMEG-1 product is purified N terminal GST-tagged recombinant PTPMEG-1 enzyme. This enzyme contains amino acids 423-end. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 1.09 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9106