Learn More
Eurofins Discoverx PTPMEG-2, 10 μg. (14-592)
Supplier:  Eurofins Discoverx 14592
Encompass
The PTPMEG-2 product is purified N terminal GST-tagged recombinant PTPMEG-2 enzyme. This enzyme contains amino acids 283-end. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg of 0.466 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9017