Learn More
Aviva Systems Biology Corp PTPN1 Antibody - middle region (ARP45360_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45360_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTPN1. Alternate names: PTP1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59012