Learn More
Sino Biological Recombinant Human PTP1B Protein (Full Length, GST Tag), Active
Supplier:  Sino Biological P2820G500
Encompass_Preferred
Recombinant full length human PTPN1 was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 50-257-3778