Learn More
Sino Biological Recombinant Human SHP-1 Protein (Full Length, His Tag), Active
Catalog No. 502573728
Encompass_Preferred
Recombinant full length human PTPN6 was expressed in E.coli cells using a C-terminal His tag.
Catalog No. 50-257-3728 Supplier Sino Biological Supplier No. P3320H500
Recombinant full length human PTPN6 was expressed in E.coli cells using a C-terminal His tag.