Learn More
Aviva Systems Biology Corp PTPN2 Antibody - middle region (ARP45363_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45363_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPN2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PTPN2. Alternate names: PTPT, TC-PTP, TCELLPTP, TCPTP, PTN2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59014