Learn More
Sino Biological PTPN2 / TC-PTP Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10570T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PTPN2 / TC-PTP (rh PTPN2 / TC-PTP; Catalog#10570-H20B; P17706-1; Met1-Asn314). PTPN2 / TC-PTP specific IgG was purified by Human PTPN2 / TC-PTP affinity chromatography.
Catalog No. 50-224-8309