Learn More
ABclonal Technology PTPN22 Rabbit pAb
SDP
Catalog No. 5000109015
Encompass_Preferred
This gene encodes of member of the non-receptor class 4 subfamily of the protein-tyrosine phosphatase family. The encoded protein is a lymphoid-specific intracellular phosphatase that associates with the molecular adapter protein CBL and may be involved in regulating CBL function in the T-cell receptor signaling pathway. Mutations in this gene may be associated with a range of autoimmune disorders including Type 1 Diabetes, rheumatoid arthritis, systemic lupus erythematosus and Graves disease. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Catalog No. 50-001-09015 Supplier ABclonal Technology Supplier No. A1406500UL
This gene encodes of member of the non-receptor class 4 subfamily of the protein-tyrosine phosphatase family. The encoded protein is a lymphoid-specific intracellular phosphatase that associates with the molecular adapter protein CBL and may be involved in regulating CBL function in the T-cell receptor signaling pathway. Mutations in this gene may be associated with a range of autoimmune disorders including Type 1 Diabetes, rheumatoid arthritis, systemic lupus erythematosus and Graves disease. Alternatively spliced transcript variants encoding distinct isoforms have been described.