Learn More
Aviva Systems Biology Corp PTPN6 Antibody - middle region (ARP61398_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61398_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPN6. It was validated on Western Blot. . Alternate names: HCP, HCPH, HPTP1C, PTP-1C, SH-PTP1, SHP-1, SHP-1L, SHP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-364