Learn More
ACROBiosystems ACROBiosystems Human PTPRD Protein, His Tag
Catalog No. 501957379
Encompass_Preferred
Recombinant Protein;1MG;Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Glu 1265 (Accession # P23468-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 140.0 kDa. The protein migrates as 19 kDa and 120-130 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7379 Supplier ACROBiosystems Supplier No. PTDH52H91MG
Recombinant Protein;1MG;Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Glu 1265 (Accession # P23468-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 140.0 kDa. The protein migrates as 19 kDa and 120-130 kDa under reducing (R) condition (SDS-PAGE).