Learn More
Aviva Systems Biology Corp PTPRE Antibody - middle region (ARP46732_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46732_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPRE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTPRE. Alternate names: DKFZp313F1310, HPTPE, PTPE, R-PTP-EPSILONApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59386