Learn More
Aviva Systems Biology Corp PTPRH Antibody - middle region (ARP43522_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43522_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPRH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTPRH. Alternate names: FLJ39938, MGC133058, MGC133059, SAP-1, SAP1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58559