Learn More
Aviva Systems Biology Corp PTPRN Antibody - middle region (ARP45381_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45381_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPRN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTPRN. Alternate names: FLJ16131, IA-2, IA-2/PTP, IA2, ICA512, R-PTP-NApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59016