Learn More
Aviva Systems Biology Corp PTPRR Antibody - N-terminal region (ARP45385_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPRR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PTPRR. Alternate names: DKFZp781C1038, EC-PTP, FLJ34328, MGC131968, MGC148170, PCPTP1, PTP-SL, PTPBR7, PTPRQApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59017