Learn More
Aviva Systems Biology Corp PTPRT Antibody - C-terminal region (ARP63448_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPRT. It was validated on Western Blot. . Alternate names: KIAA0283, RPTPrhoApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-971