Learn More
Aviva Systems Biology Corp CAVIN1 Antibody - middle region (ARP36965_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36965_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PTRF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse PTRF. Alternate names: Cavin, Cav-p60, AW546441, 2310075E07RikApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56298